Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD Microtainer™ Contact Activated Lancet
Contact Activated Lancet activates only when positioned and pressed against the skin, facilitating a consistent puncture depth for easier sampling
Supplier: BD 366576
This item is not returnable.
View return policy
Description
- The lancet enhances user comfort with an ergonomic design for a more comfortable grip
- It activates only when positioned and pressed against the skin, facilitating a consistent puncture depth for easier sampling.
- The lancet covers only a small area at the contact point, which improves the visibility of the desired puncture site.
- The intuitive procedure reduces training time, and the design prevents product reuse, reducing the possibility of patient, clinician, and/or sample contamination.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Yes | |
Pressure activated finger lancet | |
Low | |
Purple |
1.5 mm | |
Needle, 30G x 1.5mm depth | |
30 G | |
Lancet |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction