Learn More
Perkin Elmer US LLC Micro-Channel Kit for Clarus with PPC
Supplier:  Perkin Elmer US LLC N6520270
Encompass_Preferred
Request Bulk Order
Swafer micro-channel wafer technology is an innovative and user-friendly approach for flowswitching and splitting applications - it delivers unparalleled hardware and application flexibility, expanding the capabilities of capillary gas chromatography (GC).
Catalog No. 50-211-7117
Explore available promotions