Learn More
Perkin Elmer US LLC Micro-Channel Kit for Clarus with PreVent
Supplier: Perkin Elmer US LLC N6520271
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Swafer micro-channel wafer technology is an innovative and user-friendly approach for flowswitching and splitting applications - it delivers unparalleled hardware and application flexibility, expanding the capabilities of capillary gas chromatography (GC).