Learn More
World Precision Instrument Micro Ophthalmic Sterilization Baskets with Lids-WP-22000
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: World Precision Instrument WP9022000
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
- Designed specifically for delicate micro instruments
- Silicon inserts on bottom baskets to accommodate micro instruments
- Lids are provided with silicon strips to grip the instruments
- Side walls are made of perforated sheets
- User friendly locking handle
- Manufactured with stainless steel 304
- Electrolyte polished