Learn More
TARGETMOL CHEMICALS INC MG-101 25MG
Supplier: TARGETMOL CHEMICALS INC T6583
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. MG-101 (Calpain inhibitor I) is a cell-permeable and potent inhibitor of cysteine proteases including calpains and lysosomal cathepsins. purity: 98%