Learn More
Cole-Parmer Extech 407730 Digital Sound Level Meter; 40 to 130 db
Supplier: Cole-Parmer 4042532
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
Economical meter measures sound decibel levels for a variety of workplace situations including workplace machinery monitoring and audio/alarm system checks. Meter features a 40 to 130 dB adjustable measuring range with selectable A/C weighting and fast/slow response. Digital display includes 0.1 dB resolution plus analog bar graph for easy readability. MAX Hold function stores loudest dB level reached and auto power off conserves battery life over time.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.