Learn More
Electron Microscopy Sciences Metallurgical Mount Holder 1.5" and 40 mm, 3.2 mm x 8mm Pin
Supplier: Electron Microscopy Sciences 7594108
This set screw vice which is made from machined aluminum with stainless steel allen set screws has an open slot that is 10 mm (.394") wide x 5 mm (.197") deep. It measures 25 mm (1”) dia. x 17.5 mm (0.69”) high.
Type: 3.2 mm x 8 mm Pin