Learn More
Bellco Glass, Inc. Metal stone sparger 15l for flat or round bottom omni vessels. Sparger inserts through stainless steal headplate fitting. 4 1/2"wide x 20 1/2" long 316 l stainless steel ra 32 finish or better Stainless steel frit
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Bellco Glass, Inc. 196489158
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Metal stone sparger 15l for flat or round bottom omni vessels. Sparger inserts through stainless steal headplate fitting. 4 1/2"wide x 20 1/2" long 316 l stainless steel ra 32 finish or better Stainless steel frit