Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Meridian Bioscience™ Premier™ C. difficile Toxins A and B Microplate Assay
![SDP](/content/dam/fishersci/glyphs/sdp.png)
ELISA for the detection of C. difficile toxins A and B in stool specimens
Supplier: Meridian Bioscience™ 616096
Includes: Kits contain all assay microplates, diluents, controls, substrate, conjugate, pipets, and plate sealing film necessary to complete 96 tests.
Description
- CLIA Complexity: High
- Designed for accuracy and versatility with results within 30 minutes
- A qualitative enzyme immunoassay for the detection of the C. difficile toxins A and B directly from diluted stool samples
- Sensitivity: 94.7%
- Specificity: 97.3%
Specifications
CE Marked | |
C. difficile toxins A and B | |
Assay Microplates, Diluents, Controls, Substrate, conjugate, Pipets and Plate Sealing Film |
2°C to 8°C | |
Premier™ Toxins A and B Microplate Assay |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction