Learn More
Hach Company Mercury Scrubber
Supplier: Hach Company 2655800
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
For use with the Cold Vapor Mercury Test. Replace the Mercury Scrubber in the air trap housing at least once for every reagent set used. The mercury scrubber will capture mercury vapor if the Mercury Absorber Column is not properly activated using HgEx Reagent B and HgEx Reagent C. In addition, mercury is also captured if the capacity of the Absorber Column is exceeded. If the Mercury Scrubber has captured mercury vapor, it must be disposed of according to applicable regulations. Sorbent Cap = 500mg. 8 X 110MM, 1/pk
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.