Learn More
MCR Safety Indirect Vent Goggles
Click to view available options
Includes:
Adjustable headband
Neoprene headband
Wide elastic headband
Lens Coating:
Anti-fog, Scratch-resistant
Scratch-resistant
Description
Offered in both scratch-resistant polycarbonate lens and anti-fog lens, the Verdict is also available with either a wide elastic headband or a neoprene headband.
- Impact Resistant
- Fits over Prescription Glasses
- Optional Foam Lining
- Indirect Ventilation
- ANSI Z87+ and CSA Z94.3
Compliance
Meets ANSI Z87.1-2015, ANSI D3, ANSI D4 and CSA Z94.3 standards.
Specifications
Lens Coating | Anti-fog, Scratch-resistant |
Lens Tint | Clear |
Product Type | Safety Goggles |
Frame Color | Smoke |
Includes | Neoprene headband |
Material | Polycarbonate |
Frame Material | PVC |