Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ MaxiPost
Potassium channel modulator; exerts subtype-specific effects
$335.24 - $1405.92
Specifications
Purity | >98% |
---|---|
Product Type | MaxiPost |
Molecular Weight (g/mol) | 359.7 |
Inhibitors | MaxiPost |
Description
Potassium channel modulator. Acts as a positive modulator at neuronal Kv7 channels and calcium-activated K+ channels (BKCa) in HEK293 cells. Displays negative modulatory activity at Kv7.1 channels (Ki = 3.7 μM) and GABAA receptors. Displays anxiolytic activity in vivo.Specifications
>98% | |
MaxiPost | |
359.7 | |
MaxiPost |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title