Learn More
Hans Rudolph Inc Adapter with 22mm ID Medical Taper, will mate with 3700 or 3500 PNT (includes 2 female luer sample ports)
SDP
Supplier:  Hans Rudolph Inc 201144
Encompass_Preferred
Adapter with 22mm ID Medical Taper, will mate with 3700 or 3500 PNT (includes 2 female luer sample ports)
Catalog No. NC2120852