Learn More
Hampton Research HR4-943/Magnetic Base - Strong (9,000 gauss) - each
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Hampton Research HR4943
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
HR4-943/Magnetic Base - Strong (9,000 gauss) - each. Please note items from this supplier are non-returnable.