Learn More
Julabo USA Inc MAGIO MS-1200FW R1270 EA 115V
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Julabo USA Inc 9032728.N1.02
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
![Request Bulk Order](/content/dam/fishersci/glyphs/15395_Quote_Bulk_EN.jpg)
Water-Cooled Refrigerated/heating circulator, JULABO, MAGIO MS-1200FW 115V/60Hz, R1270 Refrigerant; -50…+200 °C; 1kW heater. Cooling: 1.26kW at 20C; 1.11kW at 0C; 640W at -20C, 390W @ -30C, 220W at -40C. Adjustable pressure/suction pump 16-31L/min, 3.5-13.3PSI. Color touchscreen UI with USB, RS232/RS485, Ethernet, MODBUS communication, USB data-logging; Pt100 port, multi-lingual, programmable. M16x1 pump fittings 8 and 12mm barbs. 5-7.5L bath tank. 13 x 18.5 x 27.6 in, 97lbs