Learn More
Electron Microscopy Sciences Magic Wand with Curved Tip And Flat, 6"
Supplier: Electron Microscopy Sciences 6332302
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Magic Wands are made from PEEK, a high performance plastic, with excellent chemical resistantance and withstands high temperatures (up to 300 degree C). Magic wand is ideal for use for electronic high-tech areas to laboratories, from preparation of samples to applying adhesives, using it as a stirring rod, or evendosing samples etc.
OAL: 6"
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.