Learn More
Neta Scientific Deuterium lamp w. RFID for DAD/MWD
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Neta Scientific 21400820
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Agilent Lamp, deuterium, long-life, with RFID tag, Agilent G1315C/D and G1365C/D diode array detectors - 58