Learn More
Zen Bio Inc LIPOLYSIS KIT NO CELLS
Supplier: Zen Bio Inc LIP1NC
Kit, Lipolysis, w/o cells; Glycerol; Without adipocytes; Storage condition: Glycerol Reagent A and Buffers: 4 deg.C; Glycerol Standard and Controls: -20 deg.C; Assay plate A (96-well); 1/KT