Learn More
Expression Systems BestBac 1.0 Linearized Baculovirus DNA
Supplier: Expression Systems 91001
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Expression Systems’ popular BestBac Linearized Baculovirus DNA backbone has been engineered to deliver highly efficient recombination with all polyhedron gene locus based transfer vectors. The resulting virus vector provides superior stability and purity compared to other systems.
• Unmatched productivity and stability for BEVS
• Improved production methodology can reduce need for plaque purification
Produces high titers of 5 E8 to 3 E9 IU/ml of recombinant baculovirus in P1 stock
• Generates P1 stock as quickly as Bac-to-Bac® and other systems
• Each BestBac vial contains enough material for a minimum of five cotransfections
• Suitable for other expression systems including BacMam™
• BestBac 1.0 is wild type at the cathepsin and chitinase (v-cath/chiA) loci, preferred for some applications.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.