Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Audit™ MicroControls™ Linearity FD D-Dimer bioMerieux VIDAS and miniVIDAS
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Assayed quality control material consisting of five levels of human plasma.
Supplier: Audit™ MicroControls™ K845M5
This item is not returnable.
View return policy
Description
- For use with bioMerieux VIDAS and miniVIDAS systems
- Five distinct levels that demonstrate linear relationship to each other for their respective analytes, reagents and instruments
- Intended to simulate human patient plasma samples for purpose of determining linearity, calibration verification and verification of reportable range for D-Dimer
- Analytes: D-Dimer
- Format: Freeze dried
- Package Size: 5 x 1mL
- Storage: Two days when stored at 2° to 8°C.
Specifications
Two days when stored at 2°C to 8°C. | |
bioMerieux VIDAS and miniVIDAS | |
5 x 1 mL | |
Freeze-dried |
D-dimer | |
Five distinct levels that demonstrate linear relationship to each other for their respective analytes, reagents and instruments | |
Human |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction