Learn More
World Precision Instrument Limited Reuse Punches Kit
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: World Precision Instrument WPKP1018
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Our reusable biopsy punches are multi-purpose sampling tools designed with a retractable cutting cannula. They are easy to operate, efficient, safe, fast and precise. Our punches consists of a razor sharp stainless steel cutting tip. It is designed to cut, retrieve and store the cored samples from a source materials. The source could be skin, tissue, gels, paper, cloth, leaves, paint chips, films or other thin, soft substrates. The biopsy punches are recommended as a DNA sampling tool and are also ideal for forensic use and laboratory sampling applications. The punches come sterile and are individually wrapped. They were sterilized using ethylene oxide.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.