Learn More
Clontech Labs 3P Lenti-X™ Packaging Single Shots (VSV-G), 16 Rxns
SDP
Supplier:  Clontech Labs 3P 631275
Encompass
Lenti-X Packaging Single Shots (VSV-G) provide an extremely simple and consistent one-step method for producing high-titer lentivirus. No additional transfection reagent is needed because Lenti-X Packaging Single Shots (VSV-G) consist of pre-aliquoted, lyophilized, single tubes of Xfect Transfection Reagent premixed with an optimized formulation of VSV-G pseudotyped Lenti-X lentiviral packaging plasmids. High-titer virus is produced by simply reconstituting this mixture with your lentiviral vector of choice in sterile water and adding it to 293T cells, e.g., Lenti-X 293T Cells (Cat. No. 632180), in a 10 cm dish. 16 Rxns
Catalog No. NC1032602
Explore available promotions