Learn More
Caron Products And Services LED Plant Growth, 2-Door, 50 cu. ft., 600 mole, 2 Tier
Supplier:  Caron Products And Services 7311502
Encompass_Preferred
Request Bulk Order
LED Plant Growth Chamber. 50 ft3 with 600 mole/m2/s of LED light, 2 tiers, temperature control, touchscreen. 10C to 45C, 208/230V, 60 Hz.
Catalog No. NC2676820