Learn More
Vector Laboratories Fluorescein Lectin Kit I
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Vector Laboratories FLK2100
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
The Fluorescein Lectin Screening Kit I provides 1 mg each of the following fluorescein-lectin conjugates: Con A, DBA, PNA, RCA120, SBA, UEA I, WGA. Selected to offer a variety of sugar specificities, each is prepared from our affinity-purified lectins and labeled using the high quality fluorescein isothiocyanate (excitation 495 nm, emission 515 nm) and conjugation procedures that ensure an optimal ratio of fluorophore to lectin. Supplied in solution essentially free of unconjugated fluorophore and preserved with sodium azide.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.