Learn More
Thermo Scientific LED Hollow Cathode Lamp for iCE Series
Supplier: Thermo Scientific 942339030821
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
Hollow cathode lamp containing magnesium emits specific wavelengths when electricity excites atoms for atomic absorption spectroscopy and functions as spectral line source allowing magnesium quantification.
Description
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)