Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ LDN 209929 dihydrochloride
Potent and selective haspin inhibitor
$327.12 - $1367.64
Specifications
Purity | >98% |
---|---|
CAS | 1784281-97-5 |
Product Type | LDN 209929 dihydrochloride |
Molecular Weight (g/mol) | 405.77 |
Inhibitors | LDN 209929 dihydrochloride |
Description
Potent and selective haspin kinase inhibitor (IC50 = 55 nM). Displays 180-fold selectivity over DYRK2.Specifications
>98% | |
LDN 209929 dihydrochloride | |
LDN 209929 dihydrochloride |
1784281-97-5 | |
405.77 |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title