Learn More
Enzo Life Sciences LDH Cytotoxicity WST Assay (500 tests)
SDP
Supplier:  Enzo Life Sciences ENZKIT1570500
Encompass_Preferred
The LDH cytotoxicity WST assay is a colorimetric assay kit used to determine cytotoxicity by measuring lactate dehydrogenase activity released from damaged cells. Applications: Colorimetric detection .
Catalog No. 50-201-3848
Explore available promotions