Learn More
Gold Biotechnology Inc LBA4404 Agrobacterium ElectroCompetent Cells 5x50 µl
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Gold Biotechnology Inc CC2205X50C
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
GoldBio’s LBA4404 ElectroCompetent Agrobacterium cells allow you to obtain high transformation efficiency in applications such as gDNA or cDNA library construction. Our LBA4404 strain harbor a rifampicin resistance (rif) gene. Furthermore, LBA4404 has an octoprine-type Ti plasmid pAL4404 without self-transport function, containing the vir genes. Our LBA4404 strain can be used in genetic transformation of tomato, tobacco and other plants.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.