Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Includes: Polarimeter, Dust cover, Manual
Description
Laxco™ POL-300 Series Automatic Polarimeter uses an LED light source to avoid frequent replacement of sodium lamps, and provides a more stable light source for increased accuracy. Its color, 8in LCD, touch panel display allows for easy operation to quickly nagivate through its various menus.
Each unit is inspected in the United States by a factory qualified technician under a ridged quality control process to insure its accuracy, and performance.
KEY FEATURES:
- 8in. color, touch screen display
- 589nm LED illumination with 100,000 bulb life
- RS232 and USB connectivity
- Stores 4000 data groups to internal memory
- Supports 100mm and 200mm glass tubes
- Single button measuring key
- Printer connectivity to print out readings for paper record keeping
- Ability to create user accounts
- Ability to create and manage measuring methods
- Ability to change daudrate (2400 to 38400)
- Ability to set screen saver (10 to 120 min or never)
- OPTIONAL: FD (food and drug administration) mode (CFR 21)
- 2-Year warranty
MEASURING MODES:
- Optical rotation
- Specific Rotation
- Concentration
- International sugar scale
DISPLAY:
- End point icon appears when measurement is complete
- Transmittance max alarm
- Temperature reading of last sample
- Current temperature reading
- Information key
Compliance
CFR 21 (optional)
Warranty and Services
2 years
Specifications
LCD Color Touchscreen | |
Polymer Prism; ±90° measuring range with a 0.001° of resolution at a ±0.01° of accuracy; Min0.5% sensitivity | |
50 Hz | |
220 V |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction