Learn More
LGC Biosearch Technologies LavaLAMP DNA Master Mix with Dye, 200 ReactionsSensitive and Fast DNA LAMP in an easy-to-use Master Mix format.
Supplier: LGC Biosearch Technologies 300671
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Sensitive and Fast DNA LAMP in an easy-to-use Master Mix format.Loop-mediated isothermal amplification (LAMP): Facilitates running amplification reactions in difficult test environments, enabling use outside of a high-tech laboratory.Master Mix Format: Simplifies reaction setup and minimizes optimization required to produce the best assay results.High Thermal Stability: Highly thermostable enzyme (up to 90 C for 5 min) enables preheating of reactions containing purified target DNA which, depending on the target, may increase assay sensitivity and decrease time to results.Elevated LAMP Reaction Temperature (68-74 C):Improves primer specificity and reduces background amplification depending on the DNA target.Both LavaLAMP DNA Master Mix kits contain: LavaLAMP DNA Master Mix, DNA Positive Control LAMP Primer Mix, and DNA Positive Control. The LavaLAMP DNA Master Mix with Dye also contains Green Fluorescent Dye for fluorescent detection of amplified DNA.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.