Learn More
Medicus Health Laptop Disposable Keyboard Skins, 12"W
Supplier: Medicus Health 5191M5
12"W Disposable Laptop Skins are non-sterile and have elastic edges to wrap around all sides of the keyboard portion of a laptop ∙Great for when you want to protect the unit from dust or debris in labs or pharmacies ∙3 different sizes to fit all laptops: 12", 14" 17+"∙Ideal for short term or one time use ∙Covers side ports, jacks and vents, not designed for long term use∙Available width: 12" wide - 5191M3, 14" wide - 5191M4, 17"+ wide - 5191M5
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.