Learn More
Electron Microscopy Sciences EMS Lacey Carbon Film 50 µm holes on 200 mesh Ni
Supplier: Electron Microscopy Sciences LC200NI
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
This carbon coated film on a broken pattern consists of woven-mesh-like holes of different sizes and shapes. This type of pattern provides support but does not interfere when observing specimen sections. All of our coated grids are optically checked followed by batch testing in the EM. Packed in grid storage box. NOTE: All of our film is laid on the shiny side of the grid.
Grid: Nickel
Hole: 50 µm
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.