Learn More
Labconco™ XPert™ Bulk Powder Filtered Stations
SDP
Designed to protect the user during powder transfer operations
Supplier:  Labconco™ 3962602
Request Bulk Order
 View more versions of this product
Catalog No. 10-304-159
Add to Cart
Description
Description
XPert Filtered Stations have an exhaust collar and require ducting to house exhaust or a remote blower.
  • Clean-Sweep™ openings and a zone-perforated rear baffle enhance horizontal laminar airflow
  • Built-in Guardian™ 1000 digital airflow monitor
  • Upper dilution air supply with bypass air
  • Upper containment sash foil and side-entry air foils
  • Stainless-steel work surface with front spill trough
  • Transfer caddy with caddy latch and positioning plate
  • Pushbutton operated electric hydraulic lift mechanism for raising the container to the optimal ergonomic height
  • 30 in. interior depth and 32 in. interior height
  • Tempered safety glass front sash and sides
  • 12 in. high sash opening; ergonomic angled sash with sealed gas-assisted springs holds sash open (28 in. loading height)
  • 99.99% efficient HEPA filter
  • True bag-in/bag-out filter disposal system
  • Prewired fluorescent light and switches for remote blower and light
  • Powder-coated steel exterior
  • 2 in. square-tube powder-coated steel base stand
  • Left-side mounted waste chute
  • Fourplex electrical receptacle and two utility ports
Order Info
For 115V 60Hz. International models are available; contact your Customer Service Representative.
Compliance
SEFA 1-2002, Modified ASHRAE 110-1995, ANSI Z9.5-1993, UL 3101-1/61010-1, CAN/CSA C22.2 No. 1010.1
Specifications
Specifications
Bulk Powder Filtered Station
SEFA 1-2002, ASHRAE 110-1995, ANSI Z9.5-1993, UL 3101-1/61010-1, CAN/CSA C22.2 No. 1010.1, CE
93.98 cm
12 in.
1075 lb.
Fluorescent
37 in.
86.8 in.
30.5 cm
182.8 cm
Product Suggestions
Product Suggestions