Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Labconco™ WaterPro™ Polishing Kits
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Combinations of purification technologies to meet your needs
$1152.00 - $1197.00
Catalog Number | Mfr. No. | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | For Use With (Equipment) | Price | Quantity & Availability | |||||
16-307-93
![]() |
Labconco™
9047400 |
For older models (manufactured before 8/1/93) WaterPro PS/UF Polishing Stations |
Pack for $1,197.00
|
|
|||||
16-307-87B
![]() |
Labconco™
9047401 |
For WaterPro PS/UF Polishing Stations (90007-00 to -03) |
Each for $1,152.00
|
|
|||||
16-307-62
![]() |
Labconco™
9007500 |
For WaterPro Work Stations (90000-00 and 90002-00) | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
- Timed dispense for unattended operation
- Automatic/intermittent recirculation through polishing loop to maintain water quality, minimize bacterial growth and reduce rinse up time
- Attractive and corrosion-resistant type 304 stainless steel front panel
- Unpigmented virgin natural polypropylene bowls have no impurities to contaminate water
- Water quality alert: LED flashes the actual megohm-cm when water quality falls below set point
- Built-in dispensing center
- Quiet 1/4 hp motor and rotary vane recirculation pump
- Dispensing valve
- Wall mountable
- TOC dump valve allows a small portion of purified water to drain during automatic recirculation to maintain low TOC levels (Analytical Instrument and Hybrid models)
- UV flush valve allows a small portion of purified water to rinse ultrafilter during automatic recirculation to extend the life of ultrafilter (Life Science and Hybrid models)
Certifications
CE marked (230V models)
Compliance
ETL listed (115V models)
Specifications
Polishing Kit |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title