Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Labconco™ Protector™ Standard Storage Cabinet
![SDP](/content/dam/fishersci/glyphs/sdp.png)
The Labconco Protector™ Standard Storage Cabinet may be used for storage of supplies and small laboratory instruments.
Supplier: Labconco™ 9900800
Includes: Four leveling feet and one 8 in. filler panel to increase cabinet depth from 22 to 30 in. .
Description
The Labconco Protector™ Standard Storage Cabinet may be used for storage of supplies and small laboratory instruments.
- Support loads up to 800 lb. (363kg)
- Durable epoxy-coated steel construction
- Attractive glacier white exterior complements laboratory casework and Labconco fume hoods
- Manual-closing, non-locking door(s)
Order Info
Work surface and interior shelf kits are sold separately
Specifications
Storage Cabinet | |
90.17 to 93.34 cm | |
Four Leveling Feet and One 8 in. D Filler Panel to Increase Cabinet Depth from 22 to 30 in. | |
Ideal for storage and venting of small laboratory instrument and mild acid and other reagent | |
Single, Left Hinge, Manual Close | |
22 in. | |
800 lb. | |
Epoxy-coated Steel |
12 in. | |
30.48 cm | |
29.48 kg | |
35.5 to 36.75 in. | |
362.8 kg | |
55.88 cm | |
65 lb. |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction