Learn More
Labconco™ Color-Smart Acid Gas Filter Cells for Paramount Filtered Enclosures
SDP
For use with Paramount™ Filtered and Duet Filtered Enclosures.
Supplier:  Labconco™ 6910300
Catalog No. 16-105-90
Add to Cart
Description
Description
Carbon-based acid gas filters for Paramount™ Duet Filtered Enclosures (pre-9/2008) and Paramount Filtered enclosures (pre-9/2008).
Specifications
Specifications
36, 72 in. W Paramount™ Filtered Enclosure
40.8 Kg
Gases from Hydrochloric, Nitric, Sulfuric and other Acids
90 lb.
Filter
Product Suggestions
Product Suggestions
Safety and Handling
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov