Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco™ CentriVap™ Solvent Systems
SDP

Combines multiple concentration components into one easy for ordering and just by adding vacuum pump it will be ready to concentrate samples.

Supplier:  Labconco™ 7995811

Request Bulk Order

 View more versions of this product

Catalog No. 16-109-101


Explore available promotions
Add to Cart

Includes: Includes: Benchtop concentrator with glass lid, cold trap, solvent insert, clear canister, 12-13mm rotor, vacuum tubing and six clamps

Description

Description

Combines multiple concentration components into one easy for ordering and just by adding vacuum pump it will be ready to concentrate samples.

  • Benchtop style
  • Lid type: glass
  • Type/Application: solvents
  • Accessories (Required): Diaphragm or Rotary Vane vacuum pump

Specifications

Specifications

CE
50 Hz
230 V 50 Hz
International - Schuko
Benchtop concentrator with Glass lid, cold trap, solvent insert, clear canister, 12 to 13mm rotor, vacuum tubing and 6 clamp
230 V
4 A
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.