Learn More
Poltex Lab Film Roll Dispenser
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Poltex Lab Film Roll Dispenser comes with slide cutter for film cutting.
Supplier: Poltex FILMHNGW
This item is not returnable.
View return policy
Description
- Super durable PETG parafilm roll holder.
- Comes with slide cutter for easy film cutting.
- Slide cutter mounts to either side for left or right side insertion depending on mounting direction.
- Mounting holes on ''roof'' as well as on back for mounting under an overhang, or on a wall or piece of equipment.
- Choose from wall mount (screws/anchors included), VHB tape mount (very high bond tape included), or magnet mount (powerful magnets included).
- See image for dimensions
Specifications
Parafilm Dispenser | |
PETG | |
Wall Mounting |