Learn More
Bellco Glass, Inc. Stainless steel stand for round bottom large flange omni vessels. Material: 316l stainless steel Dimensions: 24x14" Heavy duty stainless w/locking clamp Silicone cushion protects glass vessel Four legs provides stability
Supplier: Bellco Glass, Inc. 766900036
Stainless steel stand for round bottom large flange omni vessels. Material: 316l stainless steel Dimensions: 24x14" Heavy duty stainless w/locking clamp Silicone cushion protects glass vessel Four legs provides stability