Learn More
Cole-Parmer Kynar Gas Sampling Bag, 6" x 6", On/Off and Septum valves
Supplier: Cole-Parmer 0141050
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
All standard models have either an on/off valve (3⁄16'' nickel-plated brass hose barb) or a silicone septum in polypropylene housing; dual-valve models have both. Max temp: 160°F (71°C). Standard Kynar Bags Single-wide seam; available in 2-mil and 4-mil thick styles. Dual-Valve Kynar Bags Double-wide seamed bags have both on/off and septum valves for convenient mixing.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.