Learn More
Cole-Parmer KYNAR GAS SAMPLING BAG,12 X12
Supplier: Cole-Parmer 0141012
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Cole-Parmer Kynar Gas Sampling Bag, 12" x 12", On/Off valve. Corrosive-resistant Kynar for all your application needs. Strong seams for added durability
This item is not returnable.
View return policy