Learn More
TARGETMOL CHEMICALS INC KT-362 FUMARATE 5MG
Supplier: TARGETMOL CHEMICALS INC T27753
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. KT-362 fumarate is a novel compound that acts as an antagonist of calcium channels potassium channels and sodium channels.KT-362 fumarate causes vasodilation by affecting intracellular calcium mobilization in atrial muscle cells.KT-362 fumarate has a relaxing effect on femoral and basilar artery strips in rabbits. purity: 99%
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.