Description
Kinematic viscosity is of primary importance in the design and selection of a wide range of petroleum products. Calibrated capillary viscometers are used to measure flow under gravity or vacuum at precisely controlled temperatures.
- With Optical Flow Detection System
- Includes communication and power ports for each optical detection assembly; can utilize up to five optical assemblies (K23780 series)
- Two additional positions are available for manual viscosity measurements; all positions can be used in manual mode
- Modular design of three types of optical assemblies allows interchangeable use of any combination and size of Cannon™-Fenske, Reverse Flow, and Ubbelohde tubes in viscosity baths
- Integrated automatic timing on LCD microcomputer with optical sensors automatically measures sample efflux times
- Automatic calculation and display of results in viscosity units or seconds
- Bath Medium: water or suitable heat transfer fluid
- Thermometers sold separately
Compliance
Complies with ASTM D445, D2170, D6074, D6158; IP 71, 319; ISO 3104; DIN 51550; FTM 791-305; and NF T 60-100 specifications.
Specifications
Ambient to 150°C, sub-ambient to 10°C with external cooling | |
12 in. | |
115 V 60 Hz | |
60 Hz |
KV5000 | |
30.4 cm | |
115 V |