Learn More
Koehler™ Instrument Existent Gum Evaporation Bath
SDP
Tests for gum content formed during fuel storage and deposited on induction system surfaces, intake valves, stems and guides.
Supplier:  Koehler™ Instrument K33700
Request Bulk Order
 View more versions of this product
Catalog No. 13-626-132
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Alert:
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Description
Description
  • Electrically heated baths determine existent gum in aircraft turbine fuels by steam-jet evaporation and in motor and aviation gasolines by air-jet evaporation
  • Fully insulated, aluminum block design assures safe, efficient high temperature operation
  • Equipped with air/steam pressure regulator; stainless steel jets deliver air or steam flow to test wells
  • Microprocessor PID control provides quick temperature stabilization without overshoot
  • Overtemperature control circuit interrupts power should bath temperature exceed a programmed cut-off point
  • Dual LED displays provide actual and setpoint temperature values in °C/°F format
Compliance
Complies with ASTM D381; IP 131; ISO 6246; DIN 51784; FTM 791-3302; and NF M 07-004 specifications.
Specifications
Specifications
Superheater (0-3000w)
220/240 V
50/60 Hz
6 Unit
13.6 A
Product Suggestions
Product Suggestions