Learn More
DWK Life Sciences Kimble™ Kontes™ Round Bottom Three Neck Heavy Wall Distilling Flasks
With outer joints
Supplier:  DWK Life Sciences 6060001824
 View more versions of this product
Catalog No. K606000-1824
Add to Cart
Description
Description
  • Vertical side necks
Specifications
Specifications
6.33 in.
Borosilicate Glass
Vertical
24/40 Center, 24/40 side
160 mm
2000 mL
Standard taper outer joint
3
Round
Product Suggestions
Product Suggestions