Learn More
DWK Life Sciences Kimble™ Kontes™ NMR Pipets
Extended-tip NMR pipets for retrieving and adding sample in NMR tubes
Supplier:  DWK Life Sciences 8970850007
 View more versions of this product
Catalog No. K897085-0007
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View Alternate Product
Description
Description
Extended-tip NMR pipets for retrieving and adding sample in NMR tubes
  • Available with 7 and 9 in. (17.7 and 23cm) tips for use with 7 and 9 in. long NMR tubes
Specifications
Specifications
Borosilicate Glass
100/Cs.
Extended Tip Pipet
Product Suggestions
Product Suggestions