Learn More
Fujifilm Healthcare Americas Corp Ketone Body Calibrator 300
Supplier: Fujifilm Healthcare Americas Corp 41273791
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
The Ketone Body Calibrator 300 is intended to be used with the Autokit Total Ketone Bodies and Autokit 3-HB assays to establish points of reference that are used in the determination of values in the measurement of total Ketone Bodies and 3-HB in human serum or plasma. The Ketone Body Calibrator 300 is a 300 µmol/L Hydroxybutyric Acid solution. Package size: 4 x 5 mL bottles. This product is for Research Use Only, not for use in diagnostic procedures. Contact supplier for additional information at www.ivd.fujimed.com.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.