Learn More
Scientific Gear LLC The MKC-710B is a compact coulometric KF titrator, which can measure in the range of 1 ug to 300 mg H2O precisely in the sample of petroleum, chemical, pharmaceutical, and food industry.
SDP
Supplier:  Scientific Gear LLC MKC710B
Encompass_Preferred
Request Bulk Order
The MKC-710B is a compact Karl Fischer titrator (Coulometric method), which can measure H2O precisely in the range of 1ug to 300mgH2O in the sample of petroleum, pharmaceutical, beverage, and food industry.It can materialize shorter measurement and pre-titration by high electrolytic up to 2.6 mgH2O/min and can enable save time. The Reproducibility is within 0.3%CV (n=10)/KEM water-standard 1mg H2O. The display resolution is 0.1 µgram H2O. The required Anode solution is 100mL and Cathode solution is 5mL. Measurement data can be saved for up to 100. The RS-232C and USB interface are equipped to connect an optional printer, electronic balance, and optional data acquisition software (SOFT-CAP).
Catalog No. NC2129979