Learn More
KD Scientific Anti-Siphon Kit
For Legato™ 200
Supplier: KD Scientific 788303
Description
- Selection of accessories and replacement parts for your Legato™ Series Pumps
- Accommodates up to four 0.5 μL to 10 mL plastic or glass syringes
Specifications
For Legato™ 200 | |
Anti-Siphon Kit |