Learn More
AAPPTec Kaiser Test Kit
SDP
Supplier:  AAPPTec KGZ001
Encompass
The Kaiser Test Kit consists of: 50 mL of ninhydrin solution, and 50 mL of phenol + KCN solution. May be stored at room temperature for 6 months of greater.
Catalog No. NC1431926
Explore available promotions